Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319952.1 | Intron | NP_001306881.1 | |||
NM_024526.3 | Intron | NP_078802.2 | |||
NM_133181.3 | Intron | NP_573444.2 | |||
NM_139053.2 | Intron | NP_620641.1 | |||
XM_011542132.1 | Intron | XP_011540434.1 | |||
XM_011542133.1 | Intron | XP_011540435.1 | |||
XM_011542134.2 | Intron | XP_011540436.1 | |||
XM_011542135.2 | Intron | XP_011540437.1 | |||
XM_017002327.1 | Intron | XP_016857816.1 | |||
XM_017002328.1 | Intron | XP_016857817.1 | |||
XM_017002329.1 | Intron | XP_016857818.1 |