Product Details

SNP ID
rs7561996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69015139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGACTGTAGAGATACTATAGA[A/G]ATATGGATACTGTAGAGATAACCAG
Phenotype
MIM: 606410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANTXR1 PubMed Links

Gene Details

Gene
ANTXR1
Gene Name
anthrax toxin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018153.3 Intron NP_060623.2
NM_032208.2 Intron NP_115584.1
NM_053034.2 Intron NP_444262.1
XM_017005075.1 Intron XP_016860564.1
XM_017005076.1 Intron XP_016860565.1
XM_017005077.1 Intron XP_016860566.1

View Full Product Details