Product Details

SNP ID
rs7588515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:265104 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGCCGCCGGCCTAGGAACCATGA[A/G]GGGGAGGAGGCCAGGGACTGGGAGG
Phenotype
MIM: 171500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACP1 PubMed Links

Gene Details

Gene
ACP1
Gene Name
acid phosphatase 1, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040649.2 Intron NP_001035739.1
NM_004300.3 Intron NP_004291.1
NM_007099.3 Intron NP_009030.1
Gene
SH3YL1
Gene Name
SH3 and SYLF domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159597.2 Intron NP_001153069.1
NM_001282682.1 Intron NP_001269611.1
NM_001282687.1 Intron NP_001269616.1
NM_015677.3 Intron NP_056492.2

View Full Product Details