Product Details
- SNP ID
-
rs7577862
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:178488130 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCCACAAAAAGGAAAAAAAAAAA[A/G]GGTTGAATGTTGTCTTCTTTTGAAG
- Phenotype
-
MIM: 607062
MIM: 607774
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FKBP7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs185591978] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FKBP7
- Gene Name
- FK506 binding protein 7
There are no transcripts associated with this gene.
- Gene
- PLEKHA3
- Gene Name
- pleckstrin homology domain containing A3
View Full Product Details