Product Details

SNP ID
rs7574264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241691572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGTCAGTGGGCTGGGGCTGTCAC[A/G]ACAGACTACCTTAGACTGGGGAACT
Phenotype
MIM: 188345 MIM: 608525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTYMK PubMed Links

Gene Details

Gene
DTYMK
Gene Name
deoxythymidylate kinase
There are no transcripts associated with this gene.

Gene
ING5
Gene Name
inhibitor of growth family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005247047.2 Intron XP_005247104.1
XM_017005097.1 Intron XP_016860586.1
XM_017005098.1 Intron XP_016860587.1
XM_017005099.1 Intron XP_016860588.1
XM_017005100.1 Intron XP_016860589.1
XM_017005101.1 Intron XP_016860590.1
XM_017005102.1 Intron XP_016860591.1

View Full Product Details