Product Details

SNP ID
hCV189392134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43886559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTAGTCCTTTGTAACACCCCCCC[A/G]CTGCTGCCGAGAGGGCTAGATAAGG
Phenotype
MIM: 605460 MIM: 607544
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCG8 PubMed Links

Gene Details

Gene
ABCG8
Gene Name
ATP binding cassette subfamily G member 8
There are no transcripts associated with this gene.

Gene
LRPPRC
Gene Name
leucine rich pentatricopeptide repeat containing
There are no transcripts associated with this gene.

View Full Product Details