Product Details
- SNP ID
-
hCV189397821
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:152834723 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTGGTTCTCGAGGGCCCTAGAGG[A/C]TCCTGCTACGGACTGGTCGGCTCTG
- Phenotype
-
MIM: 601167
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
P2RY1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112887671] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- P2RY1
- Gene Name
- purinergic receptor P2Y1
There are no transcripts associated with this gene.
View Full Product Details