Product Details

SNP ID
rs7676926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127734987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATATTATTGTTTATGTTTGCCC[A/C]ATTAACTGGATTTATAGATTCTATT
Phenotype
MIM: 610796
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A31 PubMed Links

Gene Details

Gene
SLC25A31
Gene Name
solute carrier family 25 member 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318467.1 Intron NP_001305396.1
NM_031291.3 Intron NP_112581.1
XM_011532298.1 Intron XP_011530600.1

View Full Product Details