Product Details

SNP ID
rs8034819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41429590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTATAAAGCTCAGAAAGATATG[G/T]TTTCTTCTTACCACTAACCCACCGC
Phenotype
MIM: 611633
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RTF1 PubMed Links

Gene Details

Gene
RTF1
Gene Name
RTF1 homolog, Paf1/RNA polymerase II complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015138.4 Intron NP_055953.3

View Full Product Details