Product Details

SNP ID
rs974843
Assay Type
Functionally tested
NCBI dbSNP Submissions
70
Location
Chr.1:86549496 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAATCACTCTGACTGCAATATT[G/A]AGGACTGTAAAGAATTTCCACTTGT
Phenotype
MIM: 616857
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CLCA4 PubMed Links

Gene Details

Gene
CLCA4
Gene Name
chloride channel accessory 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012128.3 Intron NP_036260.2
XM_011541015.2 Intron XP_011539317.1

View Full Product Details