Product Details

SNP ID
rs199538471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125823663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAAAAAGAAGTCGAAAATGAG[A/G]ATGAAGACGATGATGACAGTGACAA
Phenotype
MIM: 611883 MIM: 606938
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCCIP PubMed Links

Gene Details

Gene
BCCIP
Gene Name
BRCA2 and CDKN1A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016567.3 129 Missense Mutation AAT,GAT N36D NP_057651.1
NM_078468.2 129 Missense Mutation AAT,GAT N36D NP_510868.1
NM_078469.2 129 Missense Mutation AAT,GAT N36D NP_510869.1
Gene
MIR4484
Gene Name
microRNA 4484
There are no transcripts associated with this gene.

Gene
UROS
Gene Name
uroporphyrinogen III synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000375.2 129 Intron NP_000366.1
NM_001324036.1 129 Intron NP_001310965.1
NM_001324037.1 129 Intron NP_001310966.1
NM_001324038.1 129 Intron NP_001310967.1
NM_001324039.1 129 Intron NP_001310968.1
XM_005270140.4 129 Intron XP_005270197.1
XM_011540127.1 129 Intron XP_011538429.1
XM_017016610.1 129 Intron XP_016872099.1
XM_017016611.1 129 Intron XP_016872100.1
XM_017016612.1 129 Intron XP_016872101.1

View Full Product Details