Product Details
- SNP ID
-
rs199812020
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:74843218 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAGCAATTGAAGGACTTGAGGAG[C/T]CGAATGGCTCCTCCCTGAAGAACAT
- Phenotype
-
MIM: 605880
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KAT6B
PubMed Links
Gene Details
- Gene
- KAT6B
- Gene Name
- lysine acetyltransferase 6B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256468.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
NP_001243397.1 |
NM_001256469.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
NP_001243398.1 |
NM_012330.3 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
NP_036462.2 |
XM_005269664.2 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_005269721.1 |
XM_017016000.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871489.1 |
XM_017016001.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871490.1 |
XM_017016002.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871491.1 |
XM_017016003.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871492.1 |
XM_017016004.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871493.1 |
XM_017016005.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871494.1 |
XM_017016006.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871495.1 |
XM_017016007.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871496.1 |
XM_017016008.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871497.1 |
XM_017016009.1 |
1062 |
Missense Mutation |
CCG,TCG |
P121S |
XP_016871498.1 |
View Full Product Details