Product Details

SNP ID
rs199765619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112283788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTTTGCAGAAGCCTCTGCAAA[A/C]TCGTGTGCTCCAAATAAAGCAGGTA
Phenotype
MIM: 602653
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TECTB PubMed Links

Gene Details

Gene
TECTB
Gene Name
tectorin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058222.2 127 Missense Mutation AAA,AAC K18N NP_478129.1
XM_017016604.1 127 Missense Mutation AAA,AAC K18N XP_016872093.1

View Full Product Details