Product Details

SNP ID
rs201160236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117136626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCGGTCCAAGTCCAGGCCCTTG[G/A]GCAGGATGATCTCTCGGATGGACCC
Phenotype
MIM: 604294
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
VAX1 PubMed Links

Gene Details

Gene
VAX1
Gene Name
ventral anterior homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112704.1 520 Missense Mutation CCC,CTC P92L NP_001106175.1
NM_199131.2 520 Missense Mutation CCC,CTC P92L NP_954582.1

View Full Product Details