Product Details

SNP ID
rs201329879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119651803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGCCCTTCTTCGTGGACCACA[A/C]CAGCCGCACCACTACGTGGAACGAC
Phenotype
MIM: 603883
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BAG3 PubMed Links

Gene Details

Gene
BAG3
Gene Name
BCL2 associated athanogene 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004281.3 434 Missense Mutation AAC,ACC N43T NP_004272.2
XM_005270287.1 434 Missense Mutation AAC,ACC N43T XP_005270344.1

View Full Product Details