Product Details
- SNP ID
-
rs201020026
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:32451715 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTCCAGCATTAACTACTAAAAAA[A/G]GACTACATAATTTACCATTATCACC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC7
PubMed Links
Gene Details
- Gene
- CCDC7
- Gene Name
- coiled-coil domain containing 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001026383.2 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
NP_001021554.1 |
NM_001321115.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
NP_001308044.1 |
NM_145023.5 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
NP_659460.3 |
XM_005252593.2 |
382 |
Intron |
|
|
XP_005252650.1 |
XM_006717505.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_006717568.1 |
XM_006717506.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_006717569.1 |
XM_011519663.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517965.1 |
XM_011519664.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517966.1 |
XM_011519666.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517968.1 |
XM_011519667.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517969.1 |
XM_011519668.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517970.1 |
XM_011519669.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517971.1 |
XM_011519670.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517972.1 |
XM_011519671.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517973.1 |
XM_011519672.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517974.1 |
XM_011519673.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517975.1 |
XM_011519674.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517976.1 |
XM_011519675.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517977.1 |
XM_011519676.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517978.1 |
XM_011519677.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517979.1 |
XM_011519678.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517980.1 |
XM_011519679.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517981.1 |
XM_011519680.2 |
382 |
Intron |
|
|
XP_011517982.1 |
XM_011519683.2 |
382 |
Intron |
|
|
XP_011517985.1 |
XM_011519684.2 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517986.1 |
XM_011519685.1 |
382 |
Intron |
|
|
XP_011517987.1 |
XM_011519686.2 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517988.1 |
XM_011519687.1 |
382 |
Intron |
|
|
XP_011517989.1 |
XM_011519688.2 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_011517990.1 |
XM_017016638.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_016872127.1 |
XM_017016639.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_016872128.1 |
XM_017016640.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_016872129.1 |
XM_017016641.1 |
382 |
Intron |
|
|
XP_016872130.1 |
XM_017016642.1 |
382 |
Intron |
|
|
XP_016872131.1 |
XM_017016643.1 |
382 |
Intron |
|
|
XP_016872132.1 |
XM_017016644.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_016872133.1 |
XM_017016645.1 |
382 |
Intron |
|
|
XP_016872134.1 |
XM_017016646.1 |
382 |
Intron |
|
|
XP_016872135.1 |
XM_017016647.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_016872136.1 |
XM_017016648.1 |
382 |
Intron |
|
|
XP_016872137.1 |
XM_017016649.1 |
382 |
Intron |
|
|
XP_016872138.1 |
XM_017016650.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_016872139.1 |
XM_017016651.1 |
382 |
Missense Mutation |
AGA,GGA |
R25G |
XP_016872140.1 |
XM_017016652.1 |
382 |
Intron |
|
|
XP_016872141.1 |
XM_017016653.1 |
382 |
Intron |
|
|
XP_016872142.1 |
- Gene
- LOC101929431
- Gene Name
- uncharacterized LOC101929431
There are no transcripts associated with this gene.
View Full Product Details