Product Details

SNP ID
rs201068859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68989034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCCTGAGGCCGAGGACGGCCCG[C/G]GTGCCGGTGACCACGCCCTGGGGCT
Phenotype
MIM: 606357 MIM: 609367
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX21 PubMed Links

Gene Details

Gene
DDX21
Gene Name
DEAD-box helicase 21
There are no transcripts associated with this gene.

Gene
KIF1BP
Gene Name
KIF1 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015634.3 314 Missense Mutation CGT,GGT R68G NP_056449.1
XM_017016067.1 314 Intron XP_016871556.1

View Full Product Details