Product Details

SNP ID
rs200494844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44911556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTGGGCTGGAATACACCTGCC[A/G]GCTCCTGGGCATCACCACTGCTGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM72 PubMed Links

Gene Details

Gene
TMEM72
Gene Name
transmembrane protein 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123376.2 241 Missense Mutation CAG,CGG Q15R NP_001116848.1
XM_011540069.2 241 Intron XP_011538371.1
XM_011540070.2 241 Intron XP_011538372.1
XM_011540071.2 241 UTR 5 XP_011538373.1
Gene
TMEM72-AS1
Gene Name
TMEM72 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details