Product Details

SNP ID
rs200285111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88669872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCTAAATATGACCTTCCAGCCA[C/T]AATCGACTTCATTGTAAAGAAAACT
Phenotype
MIM: 601980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPF PubMed Links

Gene Details

Gene
LIPF
Gene Name
lipase F, gastric type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198828.1 514 Missense Mutation ACA,ATA T120I NP_001185757.1
NM_001198829.1 514 Missense Mutation ACA,ATA T163I NP_001185758.1
NM_001198830.1 514 Missense Mutation ACA,ATA T130I NP_001185759.1
NM_004190.3 514 Missense Mutation ACA,ATA T153I NP_004181.1
XM_011540311.1 514 Missense Mutation ACA,ATA T153I XP_011538613.1

View Full Product Details