Product Details

SNP ID
rs200289184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21516067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTTTGTTGTGGAAATGCATTGT[C/T]TGTCTTTGGGCTAGGTGAAGAGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SKIDA1 PubMed Links

Gene Details

Gene
SKIDA1
Gene Name
SKI/DACH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207371.3 3115 Missense Mutation AAC,GAC N586D NP_997254.3
XM_005252450.3 3115 Missense Mutation AAC,GAC N586D XP_005252507.1
XM_011519480.2 3115 Missense Mutation AAC,GAC N586D XP_011517782.1
XM_017016225.1 3115 Missense Mutation AAC,GAC N586D XP_016871714.1
XM_017016226.1 3115 Missense Mutation AAC,GAC N586D XP_016871715.1

View Full Product Details