Product Details

SNP ID
rs200933454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117134412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCTGAGCCGAGAGCGCCCGTGG[T/C]GCAAGGCGGCAGCAGCGCAGGCAGG
Phenotype
MIM: 611171 MIM: 604294
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SHTN1 PubMed Links

Gene Details

Gene
SHTN1
Gene Name
shootin 1
There are no transcripts associated with this gene.

Gene
VAX1
Gene Name
ventral anterior homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112704.1 846 Missense Mutation ACC,GCC T201A NP_001106175.1
NM_199131.2 846 Intron NP_954582.1

View Full Product Details