Product Details

SNP ID
rs200099791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112286319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAAAATTCTCTCCCCTTTTCAG[A/G]GTGGCCACTGTTCACGTGAAGAACG
Phenotype
MIM: 602653
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TECTB PubMed Links

Gene Details

Gene
TECTB
Gene Name
tectorin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058222.2 484 Silent Mutation AGA,AGG R137R NP_478129.1
XM_017016604.1 484 Missense Mutation AGA,AGG R137R XP_016872093.1

View Full Product Details