Product Details

SNP ID
rs201466964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59793066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTGGGAGATGGAGGCGGCGTGG[A/G]CCGTTTGAATTTGTCAGGACTGTTG
Phenotype
MIM: 601985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC6 PubMed Links

Gene Details

Gene
CCDC6
Gene Name
coiled-coil domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005436.4 1508 Missense Mutation CCC,TCC P426S NP_005427.2

View Full Product Details