Product Details

SNP ID
rs201619012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100978365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGTGTCCTTATGACCCAGCCC[A/G]TGGCTTCACAGGCCTCATCATTGGT
Phenotype
MIM: 606075 MIM: 611848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf2 PubMed Links

Gene Details

Gene
C10orf2
Gene Name
chromosome 10 open reading frame 2
There are no transcripts associated with this gene.

Gene
MIR608
Gene Name
microRNA 608
There are no transcripts associated with this gene.

Gene
MRPL43
Gene Name
mitochondrial ribosomal protein L43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308396.1 1608 Intron NP_001295325.1
NM_032112.2 1608 Intron NP_115488.2
NM_176792.2 1608 UTR 3 NP_789762.1
NM_176793.1 1608 Intron NP_789763.1
NM_176794.1 1608 Intron NP_789764.1
XM_005270231.2 1608 Intron XP_005270288.1
XM_006718035.3 1608 Intron XP_006718098.1
XM_017016790.1 1608 Intron XP_016872279.1
Gene
SEMA4G
Gene Name
semaphorin 4G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203244.1 1608 Missense Mutation CAT,CGT H169R NP_001190173.1
NM_017893.3 1608 Missense Mutation CAT,CGT H169R NP_060363.2
XM_005270008.2 1608 Missense Mutation CAT,CGT H169R XP_005270065.1

View Full Product Details