Product Details

SNP ID
rs199513736
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2402689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCGTGGAGGGCGAACACGGGA[C/T]GGAGTATGACACGCTGCCTTCCGAC
Phenotype
MIM: 186845 MIM: 604600 MIM: 603852
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CD81 PubMed Links
Additional Information
For this assay, SNP(s) [rs2234278] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CD81
Gene Name
CD81 molecule
There are no transcripts associated with this gene.

Gene
TRPM5
Gene Name
transient receptor potential cation channel subfamily M member 5
There are no transcripts associated with this gene.

Gene
TSSC4
Gene Name
tumor suppressing subtransferable candidate 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297658.1 688 Missense Mutation ACG,ATG T19M NP_001284587.1
NM_001297659.1 688 Missense Mutation ACG,ATG T19M NP_001284588.1
NM_001297660.1 688 Missense Mutation ACG,ATG T19M NP_001284589.1
NM_001297661.1 688 Intron NP_001284590.1
NM_005706.3 688 Missense Mutation ACG,ATG T19M NP_005697.2
XM_006718118.2 688 Missense Mutation ACG,ATG T19M XP_006718181.1
XM_011519830.2 688 Missense Mutation ACG,ATG T19M XP_011518132.1

View Full Product Details