Product Details

SNP ID
rs200952940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56159834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGAAGGTCAATGTAAAAATCT[C/T]AATAGTGAACATCAGAGCCTGAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8K5 PubMed Links

Gene Details

Gene
OR8K5
Gene Name
olfactory receptor family 8 subfamily K member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004058.2 484 Missense Mutation AAG,GAG K162E NP_001004058.2

View Full Product Details