Product Details

SNP ID
rs200773036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74001420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTTCTTGTCTTGTTCAAAACG[A/G]TGATTCCCGTAACATCTGGACTTTC
Phenotype
MIM: 602044
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UCP3 PubMed Links

Gene Details

Gene
UCP3
Gene Name
uncoupling protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003356.3 1286 Missense Mutation CCG,TCG P311S NP_003347.1
NM_022803.2 1286 Intron NP_073714.1

View Full Product Details