Product Details

SNP ID
rs200941988
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:122115903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAATGTATTTCCTGGCCCTCT[A/C]TAGGCAACAAAGTCACCATAATTTG
Phenotype
MIM: 608853 MIM: 615965
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
BLID PubMed Links

Gene Details

Gene
BLID
Gene Name
BH3-like motif containing, cell death inducer
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001786.2 313 Missense Mutation AGA,ATA R7I NP_001001786.2
Gene
MIR100HG
Gene Name
mir-100-let-7a-2 cluster host gene
There are no transcripts associated with this gene.

View Full Product Details