Product Details

SNP ID
rs201036305
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6259973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGAGCGTGCAGGGAACCGGACC[C/T]GGGCCGGGGGCTTCCCTGTGCCGCC
Phenotype
MIM: 118445
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCKBR PubMed Links

Gene Details

Gene
CCKBR
Gene Name
cholecystokinin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318029.1 363 Silent Mutation CCC,CCT P15P NP_001304958.1
NM_176875.3 363 Silent Mutation CCC,CCT P15P NP_795344.1
XM_005253210.1 363 Silent Mutation CCC,CCT P15P XP_005253267.1
XM_017018516.1 363 Intron XP_016874005.1

View Full Product Details