Product Details

SNP ID
rs200064832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6846518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAGAAGTTGTTATCATTATCCT[A/G]CACTGTTGTGACTCCCATGTTGAAC
Phenotype
MIM: 608493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10A5 PubMed Links

Gene Details

Gene
OR10A5
Gene Name
olfactory receptor family 10 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178168.1 836 Missense Mutation TAC,TGC Y279C NP_835462.1

View Full Product Details