Product Details

SNP ID
rs200115079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55935859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTTAAGCAGGGGAGGGAGGTCA[C/T]AGAAAAAATGATTAATAACATTTTT
Phenotype
MIM: 608496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5I1 PubMed Links

Gene Details

Gene
OR5I1
Gene Name
olfactory receptor family 5 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006637.1 542 Missense Mutation TAT,TGT Y181C NP_006628.1

View Full Product Details