Product Details

SNP ID
rs200119869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13492557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTAGAGGAGCTCCAAGGGCAA[C/T]AAAATTGTGCACATCCTGCAGCTTC
Phenotype
MIM: 168450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTH PubMed Links

Gene Details

Gene
PTH
Gene Name
parathyroid hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000315.3 335 Missense Mutation ATT,GTT I66V NP_000306.1
NM_001316352.1 335 Missense Mutation ATT,GTT I98V NP_001303281.1

View Full Product Details