Product Details

SNP ID
rs200209929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61828430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCAGGGCGAGGGGGCCGCCGAG[C/T]GCGAGGTGTCGGTGCCCACCTTCAG
Phenotype
MIM: 606149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FADS2 PubMed Links

Gene Details

Gene
FADS2
Gene Name
fatty acid desaturase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281501.1 398 Intron NP_001268430.1
NM_001281502.1 398 Intron NP_001268431.1
NM_004265.3 398 Missense Mutation CGC,TGC R14C NP_004256.1

View Full Product Details