Product Details
- SNP ID
-
rs200520615
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:115178706 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGCACAGCATGGCGAACACCACC[A/C]CCGCCACGACGCCACCGATCACGGC
- Phenotype
-
MIM: 605686
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CADM1
PubMed Links
Gene Details
- Gene
- CADM1
- Gene Name
- cell adhesion molecule 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001098517.1 |
1193 |
Missense Mutation |
GGG,GTG |
G355V |
NP_001091987.1 |
NM_001301043.1 |
1193 |
Missense Mutation |
GGG,GTG |
G412V |
NP_001287972.1 |
NM_001301044.1 |
1193 |
Missense Mutation |
GGG,GTG |
G394V |
NP_001287973.1 |
NM_001301045.1 |
1193 |
Missense Mutation |
GGG,GTG |
G384V |
NP_001287974.1 |
NM_014333.3 |
1193 |
Missense Mutation |
GGG,GTG |
G383V |
NP_055148.3 |
XM_005271494.2 |
1193 |
Missense Mutation |
GGG,GTG |
G366V |
XP_005271551.1 |
XM_017017457.1 |
1193 |
Missense Mutation |
GGG,GTG |
G418V |
XP_016872946.1 |
XM_017017458.1 |
1193 |
Missense Mutation |
GGG,GTG |
G343V |
XP_016872947.1 |
XM_017017459.1 |
1193 |
Missense Mutation |
GGG,GTG |
G265V |
XP_016872948.1 |
XM_017017460.1 |
1193 |
Missense Mutation |
GGG,GTG |
G236V |
XP_016872949.1 |
XM_017017461.1 |
1193 |
Missense Mutation |
GGG,GTG |
G208V |
XP_016872950.1 |
View Full Product Details