Product Details

SNP ID
rs200528039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:314955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAAGATGGTTGGCGACGTGACC[A/G]GGGCCCAGGCCTATGCCTCCACCGC
Phenotype
MIM: 604456 MIM: 605578 MIM: 605579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFITM1 PubMed Links

Gene Details

Gene
IFITM1
Gene Name
interferon induced transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003641.3 400 Missense Mutation AGG,GGG R74G NP_003632.3
Gene
IFITM2
Gene Name
interferon induced transmembrane protein 2
There are no transcripts associated with this gene.

Gene
IFITM3
Gene Name
interferon induced transmembrane protein 3
There are no transcripts associated with this gene.

View Full Product Details