Product Details
- SNP ID
-
rs200669717
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5841377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACAGTAAGTATGAGGGATGATA[C/T]GATGTCCACAGAAGGGCAACCTTAA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR52E6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10742809] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR52E6
- Gene Name
- olfactory receptor family 52 subfamily E member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005167.1 |
521 |
Missense Mutation |
CAT,CGT |
H174R |
NP_001005167.1 |
View Full Product Details