Product Details

SNP ID
rs199788662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48245200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTTGTACACAGCAATTGTGCTG[C/G]GGAATTTCCTCATTGTGCTCACTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4X2 PubMed Links

Gene Details

Gene
OR4X2
Gene Name
olfactory receptor family 4 subfamily X member 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004727.1 97 Missense Mutation CGG,GGG R33G NP_001004727.1

View Full Product Details