Product Details

SNP ID
rs199605391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5507667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGGGCCTGAAGATTGGCTTCA[C/T]GATTCAGAAAGCCCATGGACCGCAG
Phenotype
MIM: 605473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
There are no transcripts associated with this gene.

Gene
UBQLN3
Gene Name
ubiquilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017481.2 2079 Missense Mutation NP_059509.1
XM_011520144.2 2079 Missense Mutation XP_011518446.1
XM_011520145.2 2079 Missense Mutation XP_011518447.1
Gene
UBQLNL
Gene Name
ubiquilin-like
There are no transcripts associated with this gene.

View Full Product Details