Product Details

SNP ID
rs199631140
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123977113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCTTTAGGACAGGGGGTATGTC[A/G]CAGAAGAAGTAGGCAATGTGGCAAG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR10S1 PubMed Links

Gene Details

Gene
OR10S1
Gene Name
olfactory receptor family 10 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004474.1 579 Silent Mutation NP_001004474.1

View Full Product Details