Product Details

SNP ID
rs199703792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56641698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGACAGCCATGAGGTAAGAGG[C/T]ACAGGTGGAGAAGGCTTTGTGCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5AP2 PubMed Links

Gene Details

Gene
OR5AP2
Gene Name
olfactory receptor family 5 subfamily AP member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002925.1 742 Missense Mutation NP_001002925.1

View Full Product Details