Product Details

SNP ID
rs199801193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69810621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCAGGCGTACTAGACACCGTC[C/T]GGTACAGCCGGGAGGCATACGTATT
Phenotype
MIM: 164950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF3 PubMed Links

Gene Details

Gene
FGF3
Gene Name
fibroblast growth factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005247.2 895 Missense Mutation CAG,CGG Q135R NP_005238.1

View Full Product Details