Product Details

SNP ID
rs199759649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35432666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACCACACTGACCACCCCAGAG[C/T]GCAGTGTGTCGTTCTTGAAGCCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAMR1 PubMed Links

Gene Details

Gene
PAMR1
Gene Name
peptidase domain containing associated with muscle regeneration 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001991.2 2350 Missense Mutation CAC,CGC H618R NP_001001991.1
NM_001282675.1 2350 Missense Mutation CAC,CGC H578R NP_001269604.1
NM_001282676.1 2350 Missense Mutation CAC,CGC H507R NP_001269605.1
NM_015430.3 2350 Missense Mutation CAC,CGC H635R NP_056245.2

View Full Product Details