Product Details
- SNP ID
-
rs202221368
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:63999755 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGTAGGCGATGGGCCCCACTGTGT[A/G]GATGACGTCTACGGGGGGCGACGGG
- Phenotype
-
MIM: 610400
MIM: 608337
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MACROD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11600062] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MACROD1
- Gene Name
- MACRO domain containing 1
- Gene
- OTUB1
- Gene Name
- OTU deubiquitinase, ubiquitin aldehyde binding 1
There are no transcripts associated with this gene.
View Full Product Details