Product Details

SNP ID
rs201468587
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:109424047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGAGAGGCAGCCCCGGGACTC[C/T]CCCTTCTGCGCCCCTTCCAACGCCT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C11orf87 PubMed Links

Gene Details

Gene
C11orf87
Gene Name
chromosome 11 open reading frame 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207645.3 921 Silent Mutation TCC,TCT S138S NP_997528.2
XM_011542817.2 921 Silent Mutation TCC,TCT S285S XP_011541119.1
XM_011542818.2 921 Silent Mutation TCC,TCT S138S XP_011541120.1

View Full Product Details