Product Details

SNP ID
rs201450116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112025514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGACGGGCTCAGAATGTAGCCCC[A/G]TGGGCGGGACTCGAGGCTCGGTGGA
Phenotype
MIM: 610493 MIM: 608770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIXDC1 PubMed Links

Gene Details

Gene
DIXDC1
Gene Name
DIX domain containing 1
There are no transcripts associated with this gene.

Gene
DLAT
Gene Name
dihydrolipoamide S-acetyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001931.4 701 Silent Mutation CCA,CCG P14P NP_001922.2
XM_011542647.2 701 Silent Mutation CCA,CCG P14P XP_011540949.1

View Full Product Details