Product Details

SNP ID
rs201806866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2402815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGCCTGGGGAGGAGGATTCAG[A/G]TCCTGATGAGCCGCCCTCACCCCCG
Phenotype
MIM: 186845 MIM: 604600 MIM: 603852
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD81 PubMed Links

Gene Details

Gene
CD81
Gene Name
CD81 molecule
There are no transcripts associated with this gene.

Gene
TRPM5
Gene Name
transient receptor potential cation channel subfamily M member 5
There are no transcripts associated with this gene.

Gene
TSSC4
Gene Name
tumor suppressing subtransferable candidate 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297658.1 814 Missense Mutation GAT,GGT D61G NP_001284587.1
NM_001297659.1 814 Missense Mutation GAT,GGT D61G NP_001284588.1
NM_001297660.1 814 Missense Mutation GAT,GGT D61G NP_001284589.1
NM_001297661.1 814 Intron NP_001284590.1
NM_005706.3 814 Missense Mutation GAT,GGT D61G NP_005697.2
XM_006718118.2 814 Missense Mutation GAT,GGT D61G XP_006718181.1
XM_011519830.2 814 Missense Mutation GAT,GGT D61G XP_011518132.1

View Full Product Details