Product Details

SNP ID
rs201810252
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.11:5788050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTAAAGAAGGTAAAGAAGGCT[G/T]GAACATAGGTGAGGACTATGGCACA
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
OR52N1 PubMed Links

Gene Details

Gene
OR52N1
Gene Name
olfactory receptor family 52 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001913.1 767 Missense Mutation CAA,CCA Q256P NP_001001913.1
Gene
OR52N5
Gene Name
olfactory receptor family 52 subfamily N member 5
There are no transcripts associated with this gene.

View Full Product Details