Product Details

SNP ID
rs202101200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17720007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGGTGCACCCGGCCCCGGGCGC[A/C]CGTGAGGACGAGCATGTGCGCGCGC
Phenotype
MIM: 159970
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYOD1 PubMed Links

Gene Details

Gene
MYOD1
Gene Name
myogenic differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002478.4 445 Silent Mutation GCA,GCC A75A NP_002469.2

View Full Product Details