Product Details

SNP ID
rs201940142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77227870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAGGCCTCTGACTTTACCTGTGC[A/G]AGTGCTGGAGGTTTCAAACAATTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDPD4 PubMed Links

Gene Details

Gene
GDPD4
Gene Name
glycerophosphodiester phosphodiesterase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182833.1 3069 Missense Mutation CGC,TGC R507C NP_878253.1
XM_011544826.1 3069 Missense Mutation CGC,TGC R309C XP_011543128.1
XM_011544833.1 3069 Missense Mutation CGC,TGC R309C XP_011543135.1
XM_011544834.1 3069 Missense Mutation CGC,TGC R533C XP_011543136.1
XM_017017348.1 3069 Missense Mutation CGC,TGC R309C XP_016872837.1
XM_017017349.1 3069 Missense Mutation CGC,TGC R309C XP_016872838.1
XM_017017350.1 3069 Missense Mutation CGC,TGC R309C XP_016872839.1
XM_017017351.1 3069 Missense Mutation CGC,TGC R309C XP_016872840.1

View Full Product Details