Product Details

SNP ID
hCV189828695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111925975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCGCAGGGCCATGAAACAACA[G/T]GACATACGTATGAACGGGTGTTACA
Phenotype
MIM: 610493
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C11orf52 PubMed Links

Gene Details

Gene
C11orf52
Gene Name
chromosome 11 open reading frame 52
There are no transcripts associated with this gene.

Gene
DIXDC1
Gene Name
DIX domain containing 1
There are no transcripts associated with this gene.

Gene
HSPB2-C11orf52
Gene Name
HSPB2-C11orf52 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details